missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Dimethylaminocinnamaldehyde, 98%

Catalog No. AC115790050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115790050 5 g Glass bottle
AC115790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115790050 Supplier Thermo Scientific Chemicals Supplier No. 115790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6203-18-5 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6203-18-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00007002
InChI Key RUKJCCIJLIMGEP-ONEGZZNKSA-N
Synonym 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent
PubChem CID 5284506
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enal
SMILES CN(C)C1=CC=C(C=C1)C=CC=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 137°C to 142°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CH=CHCHO
Quantity 5 g
Beilstein 14, 71
Formula Weight 175.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Dimethylaminocinnamaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 228-267-0
TSCA TSCA

RUO – Research Use Only