missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobutyric acid hydrochloride, 98%

Catalog No. AAA1661006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1661006 5 g
AAA1661014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1661006 Supplier Thermo Scientific Chemicals Supplier No. A1661006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69954-66-1 | C6H14ClNO2 | 167.633 g/mol

4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water and oxidizing agents.

Chemical Identifiers

CAS 69954-66-1
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
MDL Number MFCD00012613
InChI Key RDTALXUBMCLWBB-UHFFFAOYSA-N
Synonym 4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt
PubChem CID 12691640
IUPAC Name 4-(dimethylamino)butanoic acid;hydrochloride
SMILES CN(C)CCCC(=O)O.Cl

Specifications

Melting Point 153°C to 155°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 167.64
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobutyric acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only