missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylaminobenzoyl chloride, 97%
Description
4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Reacts with water.
Notes
It is sensitive to moisture. Handle and store under an inert atmosphere. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and moisture sensitive.
4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Reacts with water.
Notes
It is sensitive to moisture. Handle and store under an inert atmosphere. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and moisture sensitive.
Specifications
Specifications
| Melting Point | 142°C to 148°C |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 1100858 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 183.64 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Dimethylaminobenzoyl chloride |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only