missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzoyl chloride, 97%

Catalog No. AAL1417614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1417614 25 g
AAL1417606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1417614 Supplier Thermo Scientific Chemicals Supplier No. L1417614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4755-50-4 | C9H10ClNO | 183.635 g/mol

4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Reacts with water.

Notes
It is sensitive to moisture. Handle and store under an inert atmosphere. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and moisture sensitive.

Chemical Identifiers

CAS 4755-50-4
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.635
MDL Number MFCD00236039
InChI Key UGJDXRVQCYBXAJ-UHFFFAOYSA-N
Synonym 4-dimethylamino benzoyl chloride, benzoyl chloride, 4-dimethylamino, 4-dimethylamino benzoylchloride, 4-dimethylaminobenzoylchloride, acmc-20amjv, p-dimethylaminobenzoylchlorid, 4-dimethyaminobenzoyl chloride, p-dimethylaminobenzoyl chloride, 4-dimethlamino benzoyl chloride
PubChem CID 4400782
IUPAC Name 4-(dimethylamino)benzoyl chloride
SMILES CN(C)C1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point 142°C to 148°C
Quantity 25 g
UN Number UN3261
Beilstein 1100858
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminobenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only