missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzonitrile, 98%

CAS: 1197-19-9 | C9H10N2 | 146.193 g/mol

Supplier:  Thermo Scientific Chemicals A1311714

 View more versions of this product

Catalog No. AAA1311714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Dimethylaminobenzonitrile
72°C to 76°C
C9H10N2
25 g
971606
JYMNQRQQBJIMCV-UHFFFAOYSA-N
4-(dimethylamino)benzonitrile
70967
98%
1197-19-9
317°C to 319°C
MFCD00001815
UN3439
4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
CN(C)C1=CC=C(C=C1)C#N
146.193
146.19

Chemical Identifiers

1197-19-9
146.193
JYMNQRQQBJIMCV-UHFFFAOYSA-N
70967
CN(C)C1=CC=C(C=C1)C#N
C9H10N2
MFCD00001815
4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
4-(dimethylamino)benzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 214-819-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only