missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzonitrile, 98%

Catalog No. AAA1311706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1311706 5 g
AAA1311714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1311706 Supplier Thermo Scientific Chemicals Supplier No. A1311706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1197-19-9 | C9H10N2 | 146.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1197-19-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00001815
InChI Key JYMNQRQQBJIMCV-UHFFFAOYSA-N
Synonym 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
PubChem CID 70967
IUPAC Name 4-(dimethylamino)benzonitrile
SMILES CN(C)C1=CC=C(C=C1)C#N

Specifications

Melting Point 72°C to 76°C
Boiling Point 317°C to 319°C
Quantity 5 g
UN Number UN3439
Beilstein 971606
Formula Weight 146.19
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobenzonitrile
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 214-819-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only