missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoazobenzene-4'-carboxylic Acid 97.0+%, TCI America™

Catalog No. p-7132161
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M04235G 5 g
M042325G 25 g
2 options

Catalog No. M04235G

Supplier: TCI America M04235G

Only null left

Chemical Identifiers

CAS 6268-49-1
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00058988
InChI Key WCKQPPQRFNHPRJ-UHFFFAOYSA-N
Synonym p-Methyl Red, Para Methyl Red
PubChem CID 22650
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(O)=O

Specifications

Color Red-Yellow
Quantity 5 g
Formula Weight 269.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminoazobenzene-4′-carboxylic Acid