missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoazobenzene-4'-carboxylic Acid 97.0+%, TCI America™

Catalog No. M04235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04235G 5 g
M042325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M04235G Supplier TCI America Supplier No. M04235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6268-49-1
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00058988
InChI Key WCKQPPQRFNHPRJ-UHFFFAOYSA-N
Synonym p-Methyl Red, Para Methyl Red
PubChem CID 22650
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(O)=O

Specifications

Color Red-Yellow
Quantity 5 g
Formula Weight 269.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminoazobenzene-4′-carboxylic Acid