missing translation for 'onlineSavingsMsg'
Learn More

4'-Dimethylaminoacetophenone 98.0+%, TCI America™

Catalog No. p-7126800
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D15755G 5 g
D157525G 25 g
2 options

Catalog No. D15755G

Supplier: TCI America D15755G

Only null left

Chemical Identifiers

CAS 2124-31-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00017246
InChI Key HUDYANRNMZDQGA-UHFFFAOYSA-N
PubChem CID 75037
IUPAC Name 1-[4-(dimethylamino)phenyl]ethan-1-one
SMILES CN(C)C1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Dimethylaminoacetophenone
EINECSNumber (3)-2939
TSCA Yes