missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)phenylboronic acid pinacol ester, 97%

Catalog No. AC444960050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444960050 5 g
AC444960010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444960050 Supplier Thermo Scientific Chemicals Supplier No. 444960050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171364-78-6 | C14H22BNO2 | 247.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 171364-78-6
Molecular Formula C14H22BNO2
Molecular Weight (g/mol) 247.15
MDL Number MFCD05663854
InChI Key DGMLJJIUOFKPKB-UHFFFAOYSA-N
Synonym n,n-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-n,n-dimethylamino phenylboronic acid, pinacol ester, dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, 4-dimethylamino phenylboronic acid pinacol ester, 4-n,n-dimethylamino phenylboronic acid pinacol ester, 2-4-dimethylamino phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n,n-dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, n,n-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, acmc-209e2t, 4-n,n-dimethylamino phenylboronic acid,pinacol ester
PubChem CID 2758659
IUPAC Name N,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CN(C)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 124°C to 126°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 247.14
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)phenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only