missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)phenethyl alcohol, 99%, Thermo Scientific™

Catalog No. p-4228615
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210930010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC210930010 Supplier Thermo Scientific Chemicals Supplier No. 210930010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 50438-75-0
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.23
InChI Key CDTPAAZQBPSVGS-UHFFFAOYSA-N
Synonym 2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol
PubChem CID 93252
IUPAC Name 2-[4-(dimethylamino)phenyl]ethanol
SMILES CN(C)C1=CC=C(C=C1)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Boiling Point 116.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CH2CH2OH
Quantity 1g
Formula Weight 165.23
Percent Purity 99%
Chemical Name or Material 4-(Dimethylamino)phenethyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only