missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-(Dimethylamino)benzylidene-4-nitroaniline, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 269.30 |
| Chemical Name or Material | 4′-(Dimethylamino)benzylidene-4-nitroaniline |
Safety and Handling
| TSCA | No |