missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)benzonitrile 98.0+%, TCI America™
Supplier: TCI America D324725G
Specifications
| 4-(Dimethylamino)benzonitrile | |
| 75°C | |
| C9H10N2 | |
| 3439 | |
| JYMNQRQQBJIMCV-UHFFFAOYSA-N | |
| 4-(dimethylamino)benzonitrile | |
| 70967 | |
| ≥98.0% (GC,T) |
| 1197-19-9 | |
| 318°C | |
| 25 g | |
| 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline | |
| CN(C)C1=CC=C(C=C1)C#N | |
| 146.193 | |
| 146.19 | |
| Crystalline Powder |
Chemical Identifiers
| 1197-19-9 | |
| 146.193 | |
| 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline | |
| 4-(dimethylamino)benzonitrile |
| C9H10N2 | |
| JYMNQRQQBJIMCV-UHFFFAOYSA-N | |
| 70967 | |
| CN(C)C1=CC=C(C=C1)C#N |
Safety and Handling
TSCA : No