missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzonitrile 98.0+%, TCI America™

Catalog No. D32475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32475G 5 g
D324725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32475G Supplier TCI America Supplier No. D32475G

Chemical Identifiers

CAS 1197-19-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
InChI Key JYMNQRQQBJIMCV-UHFFFAOYSA-N
Synonym 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
PubChem CID 70967
IUPAC Name 4-(dimethylamino)benzonitrile
SMILES CN(C)C1=CC=C(C=C1)C#N

Specifications

Melting Point 75°C
Boiling Point 318°C
Quantity 5 g
UN Number 3439
Formula Weight 146.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)benzonitrile