missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzonitrile, 95%

CAS: 1197-19-9 | C9H10N2 | 146.19 g/mol

$70.57 - $273.54

Chemical Identifiers

CAS 1197-19-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001815
InChI Key JYMNQRQQBJIMCV-UHFFFAOYSA-N
Synonym 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
PubChem CID 70967
IUPAC Name 4-(dimethylamino)benzonitrile
SMILES CN(C)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115780050
SDS View Documents
Thermo Scientific Chemicals
115780050
5 g Glass bottle
Each for $70.57
Only null left
 
AC115780250
SDS View Documents
Thermo Scientific Chemicals
115780250
25 g Glass bottle
Each for $273.54
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1197-19-9
146.19
JYMNQRQQBJIMCV-UHFFFAOYSA-N
70967
CN(C)C1=CC=C(C=C1)C#N
C9H10N2
MFCD00001815
4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
4-(dimethylamino)benzonitrile

Specifications

1197-19-9
100.0
Brown
Authentic
Glass bottle
(CH3)2NC6H4CN
5 g
4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
CN(C)C1=CC=C(C=C1)C#N
146.19
146.19
Crystalline Powder
94.0
70.0°C to 76.0°C
318.0°C
95%
C9H10N2
MFCD00001815
14, 428
JYMNQRQQBJIMCV-UHFFFAOYSA-N
4-(dimethylamino)benzonitrile
70967
95%
4-(Dimethylamino)benzonitrile, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get

GHS Signal Word: Warning

EINECSNumber : 214-819-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only