Learn More
4-(Dimethylamino)antipyrine, 98+%
CAS: 58-15-1 | C13H17N3O | 231.30 g/mol
$114.03 - $1177.45
Chemical Identifiers
| CAS | 58-15-1 |
|---|---|
| Molecular Formula | C13H17N3O |
| Molecular Weight (g/mol) | 231.30 |
| MDL Number | MFCD00003142 |
| InChI Key | RMMXTBMQSGEXHJ-UHFFFAOYSA-N |
| Synonym | aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone |
| PubChem CID | 6009 |
| ChEBI | CHEBI:160246 |
| IUPAC Name | 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one |
| SMILES | CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2143618
|
Thermo Scientific Chemicals
B2143618 |
50 g |
Each for $114.03
|
|
|||||
|
AAB2143630
|
Thermo Scientific Chemicals
B2143630 |
250 g |
Each for $397.19
|
|
|||||
|
AAB214360B
|
Thermo Scientific Chemicals
B214360B |
1000 g |
Each for $1,177.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 58-15-1 | |
| 231.30 | |
| RMMXTBMQSGEXHJ-UHFFFAOYSA-N | |
| 6009 | |
| 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one |
| C13H17N3O | |
| MFCD00003142 | |
| aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone | |
| CHEBI:160246 | |
| CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1 |
Specifications
| 58-15-1 | |
| Odorless | |
| MFCD00003142 | |
| UN2811 | |
| aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone | |
| CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1 | |
| 231.30 | |
| CHEBI:160246 | |
| ≥98% |
| 105°C to 109°C | |
| C13H17N3O | |
| 50 g | |
| 14,474 | |
| RMMXTBMQSGEXHJ-UHFFFAOYSA-N | |
| 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one | |
| 6009 | |
| 231.3 | |
| 4-(Dimethylamino)antipyrine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-365-8
RTECSNumber : CD2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only