missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylamino-3-methyl-1,2-diphenylbutan-2-ol hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 2214-28-0 |
| Flash Point | Not Applicable |
| Molecular Formula | C19H25NO · HCl |
| Linear Formula | C19H25NO · HCl |
| Quantity | 25 mg |
| Formula Weight | 319.87 |
| Grade | British Pharmacopoeia (BP) Reference Standard |