missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one, 97%, Thermo Scientific™

Catalog No. RF03845EA
Change view
Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RF03845EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF03845EA Supplier Thermo Scientific Chemicals Supplier No. RF03845EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67751-23-9
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
InChI Key DFZIBCAWOSFLFR-AATRIKPKSA-N
Synonym 4-dimethylamino-1,1-dimethoxybut-3-en-2-one, e-4-dimethylamino-1,1-dimethoxybut-3-en-2-one, 1,1-dimethoxy-4-dimethylaminobut-3-en-2-one, 1e-4,4-dimethoxy-3-oxobut-1-en-1-yl dimethylamine, 3e-4-dimethylamino-1,1-dimethoxybut-3-en-2-one, e-1,1-dimethoxy-4-dimethylaminobut-3-en-2-one, hcvl@nbfryyuffjjafp, e-4-dimethylamino-1,1-dimethoxy-but-3-en-2, 4-dimethylamino-1,1-dimethoxy-3-butene-2-one, e-4-dimethylamino-1,1-dimethoxy-but-3-en-2-one
PubChem CID 5709580
IUPAC Name (E)-4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one
SMILES CN(C)C=CC(=O)C(OC)OC

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)2NCH=CHC(O)CH(OCH3)2
Quantity 10g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)-1, 1-dimethoxybut-3-en-2-one