missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)benzaldehyde 98.0+%, TCI America™

Catalog No. p-7128229
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43341G 1 g
D43345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43341G Supplier TCI America Supplier No. D43341G
Only null left

Chemical Identifiers

CAS 73960-07-3
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042252
InChI Key ZWCXOJYJJINQGU-UHFFFAOYSA-N
Synonym 4-difluoromethoxy benzaldehyde, 4-difluoromethoxy-benzaldehyde, p-difluoromethoxy benzaldehyde, p-difluoromethoxybenzaldehyde, benzaldehyde, 4-difluoromethoxy, 1-difluoromethoxy-4-formylbenzene, pubchem7137, acmc-1bb9r, akos bb62
PubChem CID 2736986
IUPAC Name 4-(difluoromethoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC(F)F

Specifications

Color Yellow
Boiling Point 87°C
Quantity 1 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Difluoromethoxy)benzaldehyde