missing translation for 'onlineSavingsMsg'
Learn More

4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester, 96%

CAS: 1628442-55-6 | C13H16BF3O3 | 288.073 g/mol

$263.03 - $826.16

Chemical Identifiers

CAS 1628442-55-6
Molecular Formula C13H16BF3O3
Molecular Weight (g/mol) 288.073
MDL Number MFCD22419269
InChI Key LLJRSXMGPKHUQX-UHFFFAOYSA-N
Synonym 2-4-difluoromethoxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 79661309
IUPAC Name 2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)OC(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59880MD
View Documents
Thermo Scientific Chemicals
H59880MD
250 mg N/A
Only null left
 
AAH5988003
View Documents
Thermo Scientific Chemicals
H5988003
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1628442-55-6
288.073
LLJRSXMGPKHUQX-UHFFFAOYSA-N
79661309
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)OC(F)F)F
C13H16BF3O3
MFCD22419269
2-4-difluoromethoxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

1628442-55-6
MFCD22419269
2-4-difluoromethoxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)OC(F)F)F
288.073
288.07
4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
C13H16BF3O3
250 mg
LLJRSXMGPKHUQX-UHFFFAOYSA-N
2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
79661309
96%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only