missing translation for 'onlineSavingsMsg'
Learn More

4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH59880MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59880MD 250 mg
AAH5988003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59880MD Supplier Thermo Scientific Chemicals Supplier No. H59880MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1628442-55-6 | C13H16BF3O3 | 288.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1628442-55-6
Molecular Formula C13H16BF3O3
Molecular Weight (g/mol) 288.073
MDL Number MFCD22419269
InChI Key LLJRSXMGPKHUQX-UHFFFAOYSA-N
Synonym 2-4-difluoromethoxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 79661309
IUPAC Name 2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)OC(F)F)F

Specifications

Quantity 250 mg
Formula Weight 288.07
Percent Purity 96%
Chemical Name or Material 4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only