missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Diethylcarbamoyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$234.87 - $234.87
Specifications
| CAS | 389621-80-1 |
|---|---|
| Melting Point | 142°C |
| Color | White |
| Molecular Formula | C11H16BNO3 |
| MDL Number | MFCD03411949 |
Specifications
| 389621-80-1 | |
| White | |
| MFCD03411949 | |
| [4-(diethylcarbamoyl)phenyl]boronic acid | |
| 221.06 |
| 142°C | |
| C11H16BNO3 | |
| ZCGVBHIMRVYWOH-UHFFFAOYSA-N | |
| 2773375 | |
| Crystalline Powder |