missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

$285.99 - $285.99

Chemical Identifiers

CAS 389621-80-1
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD03411949
InChI Key ZCGVBHIMRVYWOH-UHFFFAOYSA-N
Synonym 4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
PubChem CID 2773375
IUPAC Name [4-(diethylcarbamoyl)phenyl]boronic acid
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5298606
View Documents
Thermo Scientific Chemicals
H5298606
5 g
Each for $285.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

389621-80-1
221.06
ZCGVBHIMRVYWOH-UHFFFAOYSA-N
2773375
CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
C11H16BNO3
MFCD03411949
4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
[4-(diethylcarbamoyl)phenyl]boronic acid

Specifications

389621-80-1
C11H16BNO3
5 g
ZCGVBHIMRVYWOH-UHFFFAOYSA-N
[4-(diethylcarbamoyl)phenyl]boronic acid
2773375
98%
138°C to 142°C
MFCD03411949
4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
221.06
221.06
4-(Diethylcarbamoyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only