missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminosalicylaldehyde, 99%

Catalog No. AAL1165906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1165906 5 g
AAL1165914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1165906 Supplier Thermo Scientific Chemicals Supplier No. L1165906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17754-90-4 | C11H15NO2 | 193.246 g/mol

4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.

Solubility
Insoluble in water.

Notes
It is sensitive to air. Store at room temperature. Incompatible with strong bases and strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 17754-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00003326
InChI Key XFVZSRRZZNLWBW-UHFFFAOYSA-N
Synonym 4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde
PubChem CID 87293
IUPAC Name 4-(diethylamino)-2-hydroxybenzaldehyde
SMILES CCN(CC)C1=CC(=C(C=C1)C=O)O

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
Beilstein 2212234
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 193.25
Percent Purity 99%
Chemical Name or Material 4-Diethylaminosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-745-3
RTECSNumber CU5615150
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only