missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)salicylaldehyde 98.0+%, TCI America™

Supplier:  TCI America D1752250G

 View more versions of this product

Catalog No. D1752250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(Diethylamino)salicylaldehyde
60°C
MFCD00003326
4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde
CCN(CC)C1=CC(=C(C=C1)C=O)O
193.246
193.25
Crystalline Powder
17754-90-4
C11H15NO2
250 g
XFVZSRRZZNLWBW-UHFFFAOYSA-N
4-(diethylamino)-2-hydroxybenzaldehyde
87293
≥98.0% (GC,T)

Chemical Identifiers

17754-90-4
193.246
XFVZSRRZZNLWBW-UHFFFAOYSA-N
87293
CCN(CC)C1=CC(=C(C=C1)C=O)O
C11H15NO2
MFCD00003326
4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde
4-(diethylamino)-2-hydroxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1191

RTECSNumber : CU5615150

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions