missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)salicylaldehyde 98.0+%, TCI America™

Catalog No. D175225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D175225G 25 g
D1752250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D175225G Supplier TCI America Supplier No. D175225G

Chemical Identifiers

CAS 17754-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00003326
InChI Key XFVZSRRZZNLWBW-UHFFFAOYSA-N
Synonym 4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde
PubChem CID 87293
IUPAC Name 4-(diethylamino)-2-hydroxybenzaldehyde
SMILES CCN(CC)C1=CC(=C(C=C1)C=O)O

Specifications

Melting Point 60°C
Quantity 25 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Diethylamino)salicylaldehyde
EINECSNumber (3)-1191
RTECSNumber CU5615150
TSCA Yes