missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)azobenzene, 98%

Catalog No. AAB2391506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2391506 5 g
AAB2391514 25 g
AAB2391522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2391506 Supplier Thermo Scientific Chemicals Supplier No. B2391506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2481-94-9 | C16H19N3 | 253.349 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2481-94-9
Molecular Formula C16H19N3
Molecular Weight (g/mol) 253.349
MDL Number MFCD00009043
InChI Key SJJISKLXUJVZOA-UHFFFAOYSA-N
Synonym 4-diethylamino azobenzene, oil yellow de, diethyl yellow, c.i. solvent yellow 56, solvent yellow 56, 4-diethylaminoazobenzene, oil yellow ga, oil yellow nb, oil yellow dea, oil yellow enc
PubChem CID 17204
IUPAC Name N,N-diethyl-4-phenyldiazenylaniline
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=CC=C2

Specifications

Melting Point 93°C to 96°C
Quantity 5 g
Formula Weight 253.35
Percent Purity 98%
Chemical Name or Material 4-(Diethylamino)azobenzene
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 219-616-8
RTECSNumber CX9871000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only