missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylamino-2-butyn-1-ol 95.0+%, TCI America™

Catalog No. p-7127510
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28055ML 5 mL
D280525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28055ML Supplier TCI America Supplier No. D28055ML
Only null left

Chemical Identifiers

CAS 10575-25-4
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00671356
InChI Key ACGZBRWTWOZSFU-UHFFFAOYSA-N
Synonym 4-diethylamino but-2-yn-1-ol, 4-diethylamino-2-butyn-1-ol, unii-ma2de62u5w, ma2de62u5w, 2-butyn-1-ol,4-diethylamino, 2-butyn-1-ol, 4-diethylamino, acmc-2098hk, 4-diethylamino-2-butyne-ol, 1-diethylaminobut-2-yn-4-ol
PubChem CID 82735
IUPAC Name 4-(diethylamino)but-2-yn-1-ol
SMILES CCN(CC)CC#CCO

Specifications

Quantity 5 mL
Formula Weight 141.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Diethylamino-2-butyn-1-ol