missing translation for 'onlineSavingsMsg'
Learn More

4-(Dibutylamino)salicylaldehyde 96.0+%, TCI America™

Catalog No. D32605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32605G 5 g
D326025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32605G Supplier TCI America Supplier No. D32605G
Only null left

Chemical Identifiers

CAS 57771-09-2
Molecular Formula C15H23NO2
Molecular Weight (g/mol) 249.354
InChI Key BWHGDPKOFVNGFQ-UHFFFAOYSA-N
Synonym 4-(Dibutylamino)-2-hydroxybenzaldehyde
PubChem CID 18534534
IUPAC Name 4-(dibutylamino)-2-hydroxybenzaldehyde
SMILES CCCCN(CCCC)C1=CC(=C(C=C1)C=O)O

Specifications

Quantity 5 g
Formula Weight 249.35
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Dibutylamino)salicylaldehyde
TSCA No
Recommended Storage Freezer