missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Diazo-N,N-dimethylaniline Chloride Zinc Chloride Hydrate 95.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| UN Number | 1325 |
| Formula Weight | 319.92 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Diazo-N,N-dimethylaniline Chloride Zinc Chloride Hydrate |
Safety and Handling
| EINECSNumber | (3)-0253 |
| TSCA | Yes |