missing translation for 'onlineSavingsMsg'
Learn More

4-Diazo-N-ethyl-N-(2-hydroxyethyl)aniline Chloride Zinc Chloride 94.0+%, TCI America™

Catalog No. D014225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D014225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D014225G Supplier TCI America Supplier No. D014225G
Only null left

Chemical Identifiers

CAS 13532-96-2
Molecular Formula C10H14Cl3N3OZn
Molecular Weight (g/mol) 363.97
MDL Number MFCD00198036
InChI Key FOZURDCUSZRTCY-UHFFFAOYSA-K
Synonym N-Ethyl-N-(2-hydroxyethyl)-1,4-phenylenediamine Diazonium Chloride Zinc Chloride
IUPAC Name zinc(2+) 4-[ethyl(2-hydroxyethyl)amino]benzene-1-diazonium trichloride
SMILES [Cl-].[Cl-].[Cl-].[Zn++].CCN(CCO)C1=CC=C(C=C1)[N+]#N

Specifications

Quantity 25 g
UN Number 1325
Formula Weight 591.66
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diazo-N-ethyl-N-(2-hydroxyethyl)aniline Chloride Zinc Chloride
EINECSNumber (3)-2560
RTECSNumber KK9240000
TSCA Yes