missing translation for 'onlineSavingsMsg'
Learn More

4-(Di-p-tolylamino)benzaldehyde 98.0+%, TCI America™

Catalog No. D27371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27371G 1 g
D27375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27371G Supplier TCI America Supplier No. D27371G

Chemical Identifiers

CAS 42906-19-4
Molecular Formula C21H19NO
Molecular Weight (g/mol) 301.389
MDL Number MFCD03093257
InChI Key XCGLXUJEPIVZJM-UHFFFAOYSA-N
PubChem CID 170701
IUPAC Name 4-(4-methyl-N-(4-methylphenyl)anilino)benzaldehyde
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C=O

Specifications

Melting Point 109°C
Quantity 1 g
Synonym 4-[Bis(4-methylphenyl)amino]benzaldehyde, 4-Formyl-4′C,4′C′C-dimethyltriphenylamine
Formula Weight 301.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Di-p-tolylamino)benzaldehyde