missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzeneboronic acid, 98%

Catalog No. AAL18076MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18076MD 250 mg
AAL1807603 1 g
AAL1807606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL18076MD Supplier Thermo Scientific Chemicals Supplier No. L18076MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 374538-04-2 | C12H17BO2 | 204.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374538-04-2
Molecular Formula C12H17BO2
Molecular Weight (g/mol) 204.076
MDL Number MFCD02093068
InChI Key KNQVRFYNQWNYPU-UHFFFAOYSA-N
Synonym 4-cyclohexylbenzeneboronic acid, 4-cyclohexylphenyl boronic acid, boronic acid, 4-cyclohexylphenyl, 4-cyclohexylphenylboronicacid, zlchem 189, pubchem6403, pubchem7791, acmc-209it1, 4-cyclohexylphenylboronic-acid, 4-cyclohexyl-phenylboronic acid
PubChem CID 4589189
IUPAC Name (4-cyclohexylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C2CCCCC2)(O)O

Specifications

Melting Point ∼220°C
Quantity 250 mg
Formula Weight 204.08
Percent Purity 98%
Chemical Name or Material 4-Cyclohexylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only