missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylaniline, 97%

Catalog No. AAL0318406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0318406 5 g
AAL0318403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0318406 Supplier Thermo Scientific Chemicals Supplier No. L0318406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6373-50-8 | C16H12Cl2N4OS | 379.26 g/mol

4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6373-50-8
Molecular Formula C16H12Cl2N4OS
Molecular Weight (g/mol) 379.26
MDL Number MFCD00001454
InChI Key YDTTUXLKBBQRHA-UHFFFAOYSA-N
Synonym benzenamine, 4-cyclohexyl, 4-cyclohexylbenzenamine, aniline, p-cyclohexyl, p-cyclohexylaniline, 4-cyclohexylphenyl amine, 4-cyclohexylphenylamine, 4-cyclohexylanilin, 4-cyclohexyl-aniline, acmc-20aivf, p-cyclohexylphenylamine
PubChem CID 80764
IUPAC Name 4-cyclohexylaniline
SMILES CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1

Specifications

Melting Point 52°C to 56°C
Boiling Point 166°C (13 mmHg)
Quantity 5 g
Beilstein 2965799
Solubility Information Insoluble in water.
Formula Weight 175.28
Percent Purity 97%
Chemical Name or Material 4-Cyclohexylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-918-9
TSCA No

RUO – Research Use Only