missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Cyclohexylacetophenone 98.0+%, TCI America™
Supplier: TCI America C121725G
Specifications
| 4′-Cyclohexylacetophenone | |
| 67°C | |
| C14H18O | |
| 25 g | |
| MSDQNIRGPBARGC-UHFFFAOYSA-N | |
| 1-(4-cyclohexylphenyl)ethan-1-one | |
| 87715 | |
| ≥98.0% (GC) |
| 18594-05-3 | |
| 128°C | |
| MFCD00001453 | |
| 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone | |
| CC(=O)C1=CC=C(C=C1)C1CCCCC1 | |
| 202.30 | |
| 202.30 | |
| Crystalline Powder |
Chemical Identifiers
| 18594-05-3 | |
| 202.30 | |
| MSDQNIRGPBARGC-UHFFFAOYSA-N | |
| 87715 | |
| CC(=O)C1=CC=C(C=C1)C1CCCCC1 |
| C14H18O | |
| MFCD00001453 | |
| 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone | |
| 1-(4-cyclohexylphenyl)ethan-1-one |
Safety and Handling
TSCA : No