missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyclohexylacetophenone 98.0+%, TCI America™

Catalog No. C12175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12175G 5 g
C121725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12175G Supplier TCI America Supplier No. C12175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18594-05-3
Molecular Formula C14H18O
Molecular Weight (g/mol) 202.30
MDL Number MFCD00001453
InChI Key MSDQNIRGPBARGC-UHFFFAOYSA-N
Synonym 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone
PubChem CID 87715
IUPAC Name 1-(4-cyclohexylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(C=C1)C1CCCCC1

Specifications

Melting Point 67°C
Boiling Point 128°C
Quantity 5 g
Formula Weight 202.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Cyclohexylacetophenone