missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Cyclohexylacetophenone, 99%
CAS: 18594-05-3 | C14H18O | 202.30 g/mol
$49.26 - $370.71
Chemical Identifiers
| CAS | 18594-05-3 |
|---|---|
| Molecular Formula | C14H18O |
| Molecular Weight (g/mol) | 202.30 |
| MDL Number | MFCD00001453 |
| InChI Key | MSDQNIRGPBARGC-UHFFFAOYSA-N |
| Synonym | 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone |
| PubChem CID | 87715 |
| SMILES | CC(=O)C1=CC=C(C=C1)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2083806
|
Thermo Scientific Chemicals
B2083806 |
5 g |
Each for $49.26
|
|
|||||
|
AAB2083814
|
Thermo Scientific Chemicals
B2083814 |
25 g |
Each for $130.79
|
|
|||||
|
AAB2083822
|
Thermo Scientific Chemicals
B2083822 |
100 g |
Each for $370.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18594-05-3 | |
| 202.30 | |
| MSDQNIRGPBARGC-UHFFFAOYSA-N | |
| 87715 |
| C14H18O | |
| MFCD00001453 | |
| 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone | |
| CC(=O)C1=CC=C(C=C1)C1CCCCC1 |
Specifications
| 18594-05-3 | |
| 179°C to 181°C (14 mmHg) | |
| MFCD00001453 | |
| 1962365 | |
| MSDQNIRGPBARGC-UHFFFAOYSA-N | |
| 1-(4-cyclohexylphenyl)ethan-1-one | |
| 87715 | |
| 99% |
| 66°C to 70°C | |
| C14H18O | |
| 5 g | |
| 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone | |
| CC(=O)C1=CC=C(C=C1)C1CCCCC1 | |
| 202.30 | |
| 202.3 | |
| 4'-Cyclohexylacetophenone |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 242-432-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only