missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyclohexylacetophenone, 99%

Catalog No. AAB2083822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2083822 100 g
AAB2083806 5 g
AAB2083814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2083822 Supplier Thermo Scientific Chemicals Supplier No. B2083822

CAS: 18594-05-3 | C14H18O | 202.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18594-05-3
Molecular Formula C14H18O
Molecular Weight (g/mol) 202.30
MDL Number MFCD00001453
InChI Key MSDQNIRGPBARGC-UHFFFAOYSA-N
Synonym 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone
PubChem CID 87715
SMILES CC(=O)C1=CC=C(C=C1)C1CCCCC1

Specifications

Melting Point 66°C to 70°C
Boiling Point 179°C to 181°C (14 mmHg)
Quantity 100 g
Beilstein 1962365
IUPAC Name 1-(4-cyclohexylphenyl)ethanone
Formula Weight 202.3
Percent Purity 99%
Chemical Name or Material 4'-Cyclohexylacetophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-432-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only