missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyclohexyl N-(tert-Butoxycarbonyl)-L-aspartate 98.0+%, TCI America™
$127.07 - $325.36
Chemical Identifiers
| CAS | 73821-95-1 |
|---|---|
| Molecular Formula | C15H25NO6 |
| Molecular Weight (g/mol) | 315.366 |
| MDL Number | MFCD00061996 |
| InChI Key | NLPQIWFEEKQBBN-NSHDSACASA-N |
| Synonym | boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester |
| PubChem CID | 7009416 |
| IUPAC Name | (2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O |
Chemical Identifiers
| 73821-95-1 | |
| 315.366 | |
| NLPQIWFEEKQBBN-NSHDSACASA-N | |
| 7009416 | |
| CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O |
| C15H25NO6 | |
| MFCD00061996 | |
| boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester | |
| (2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Specifications
| 73821-95-1 | |
| White | |
| MFCD00061996 | |
| boc-asp ochx-oh, boc-asp ochex-oh, boc-l-aspartic acid 4-cyslohexyl ester, n-boc-l-aspartic acid 4-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-asp o-chex, ambotzbaa1074, pubchem12097, 4-cyclohexyl n-boc-l-aspartate, boc-l-aspartic acid b-cyclohexyl ester | |
| CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O | |
| 315.366 | |
| 315.37 | |
| Crystalline Powder |
| 96°C | |
| C15H25NO6 | |
| 5 g | |
| NLPQIWFEEKQBBN-NSHDSACASA-N | |
| (2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
| 7009416 | |
| ≥98.0% (HPLC,T) | |
| 4-Cyclohexyl N-(tert-Butoxycarbonyl)-L-aspartate |
Safety and Handling
TSCA : No