missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanopyridine N-Oxide 98.0+%, TCI America™
$233.00 - $1985.22
Chemical Identifiers
| CAS | 14906-59-3 |
|---|---|
| Molecular Formula | C6H4N2O |
| Molecular Weight (g/mol) | 120.11 |
| MDL Number | MFCD00006205 |
| InChI Key | QNCSFBSIWVBTHE-UHFFFAOYSA-N |
| Synonym | Isonicotinonitrile N-Oxide |
| PubChem CID | 84684 |
| IUPAC Name | 4-cyanopyridin-1-ium-1-olate |
| SMILES | [O-][N+]1=CC=C(C=C1)C#N |
Chemical Identifiers
| 14906-59-3 | |
| 120.11 | |
| QNCSFBSIWVBTHE-UHFFFAOYSA-N | |
| 84684 | |
| [O-][N+]1=CC=C(C=C1)C#N |
| C6H4N2O | |
| MFCD00006205 | |
| Isonicotinonitrile N-Oxide | |
| 4-cyanopyridin-1-ium-1-olate |
Specifications
| 14906-59-3 | |
| Yellow | |
| MFCD00006205 | |
| Isonicotinonitrile N-Oxide | |
| [O-][N+]1=CC=C(C=C1)C#N | |
| 120.11 | |
| 120.11 | |
| Crystalline Powder |
| 230°C | |
| C6H4N2O | |
| 25 g | |
| QNCSFBSIWVBTHE-UHFFFAOYSA-N | |
| 4-cyanopyridin-1-ium-1-olate | |
| 84684 | |
| ≥98.0% (T) | |
| 4-Cyanopyridine N-Oxide |
Safety and Handling
TSCA : Yes