missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopyridine, 98%

Catalog No. p-7080662
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1028609 10 g
AAA1028622 100 g
AAA1028636 500 g
AAA102860E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1028609 Supplier Thermo Scientific Chemicals Supplier No. A1028609

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-48-1 | C6H4N2 | 104.112 g/mol

4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

Solubility
Soluble in water.

Notes
Incompatible with strong acids, strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 100-48-1
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.112
MDL Number MFCD00006417
InChI Key GPHQHTOMRSGBNZ-UHFFFAOYSA-N
Synonym 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
PubChem CID 7506
ChEBI CHEBI:28020
IUPAC Name pyridine-4-carbonitrile
SMILES C1=CN=CC=C1C#N

Specifications

Melting Point 76°C to 80°C
pH 6
Boiling Point 194°C to 196°C
Flash Point 88°C (190°F)
Odor Pungent
Quantity 10 g
Beilstein 107712
Solubility Information Soluble in water.
Formula Weight 104.11
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder or Lumps or Fused Solid or Solid
Chemical Name or Material 4-Cyanopyridine
Show More Show Less
Hazard Category H227-H302-H312-H315-H319-H332-H335
Hazard Statement GHS H StatementH302-H319 Harmful if swallowed. Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-856-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only