missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylhydrazine hydrochloride, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 392300010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC392300010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2863-98-1
133.15
DZUUSHCOMPROCJ-UHFFFAOYSA-N
16212962
NNC1=CC=C(C=C1)C#N
C7H7N3
MFCD00673994
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
4-hydrazinylbenzonitrile;hydrochloride

Specifications

4-Cyanophenylhydrazine hydrochloride
97.0
>110°C
97% min. (HPLC)
C7H7N3
1g
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
DZUUSHCOMPROCJ-UHFFFAOYSA-N
4-hydrazinylbenzonitrile;hydrochloride
16212962
97+%
2863-98-1
100.0
Authentic
Glass bottle
NCC6H4NHNH2·HCl
MFCD00673994
Solubility in water: soluble
NNC1=CC=C(C=C1)C#N
133.15
169.61
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions