missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylacetic acid, 97%

Catalog No. AAH2606206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2606206 5 g
AAH2606203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2606206 Supplier Thermo Scientific Chemicals Supplier No. H2606206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5462-71-5 | C9H7NO2 | 161.16 g/mol

4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Soluble in CHCl3, THF, dioxane.

Notes
Storage temperature is 2 - 4°C and Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 5462-71-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD06798066
InChI Key WEBXRQONNWEETE-UHFFFAOYSA-N
Synonym 2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
PubChem CID 79587
IUPAC Name 2-(4-cyanophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(C=C1)C#N

Specifications

Melting Point 150°C to 152°C
Quantity 5 g
UN Number UN3439
Solubility Information Soluble in CHCl3,THF,dioxane.
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 4-Cyanophenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 226-753-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only