missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl 4-n-hexylbenzoate, 99%

Catalog No. AAB2247103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2247103 1 g
AAB2247106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2247103 Supplier Thermo Scientific Chemicals Supplier No. B2247103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50793-85-6 | C20H21NO2 | 307.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50793-85-6
Molecular Formula C20H21NO2
Molecular Weight (g/mol) 307.39
MDL Number MFCD00600506
InChI Key DEUWEGPRKHPNKB-UHFFFAOYSA-N
Synonym benzoic acid, 4-hexyl-, 4-cyanophenyl ester, 4-cyanophenyl 4-n-hexylbenzoate, p-cyanophenyl p-hexylbenzoate, 4-cyanophenyl 4-hexylbenzoate, 4-hexylbenzoic acid 4-cyanophenyl, 4-n-hexylbenzoic acid 4-cyanophenyl ester
PubChem CID 170907
IUPAC Name (4-cyanophenyl) 4-hexylbenzoate
SMILES CCCCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C#N

Specifications

Melting Point 44°C to 47°C
Quantity 1 g
Formula Weight 307.39
Percent Purity 99%
Chemical Name or Material 4-Cyanophenyl 4-n-hexylbenzoate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 256-767-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only