missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl 4-Heptylbenzoate 98.0+%, TCI America™

Catalog No. h08105g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08105G 5 g
H08101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08105G Supplier TCI America Supplier No. H08105G

Chemical Identifiers

CAS 38690-76-5
Molecular Formula C21H23NO2
Molecular Weight (g/mol) 321.42
MDL Number MFCD00001817
InChI Key ZWAUTNYPKHVAEZ-UHFFFAOYSA-N
Synonym 4-Heptylbenzoic Acid 4-Cyanophenyl Ester
PubChem CID 170097
IUPAC Name (4-cyanophenyl) 4-heptylbenzoate
SMILES CCCCCCCC1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)C#N

Specifications

Melting Point 57°C
Quantity 5 g
Formula Weight 321.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanophenyl 4-Heptylbenzoate