missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanoimidazole, 95%

Catalog No. AC461360010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461360010 1 g
AC461360050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461360010 Supplier Thermo Scientific Chemicals Supplier No. 461360010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57090-88-7 | C4H3N3 | 93.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57090-88-7
Molecular Formula C4H3N3
Molecular Weight (g/mol) 93.09
MDL Number MFCD00233358
InChI Key NWVGXXPWOYZODV-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carbonitrile, 4-cyanoimidazole, 4-cyano-1h-imidazole, 3h-imidazole-4-carbonitrile, imidazole-4-carbonitrile, pubchem18770, acmc-209lvz, 5-imidazole carbonitrile, 1h-imidazol-4-kohlenitrile, ksc269a8n
PubChem CID 564457
IUPAC Name 1H-imidazole-5-carbonitrile
SMILES N#CC1=CN=CN1

Specifications

Melting Point 32.0°C to 35.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 93.09
Percent Purity 95%
Chemical Name or Material 4-Cyanoimidazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear prote

RUO – Research Use Only