missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7124841
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C11825G 5 g
C118225G 25 g
2 options

Catalog No. C11825G

Supplier: TCI America C11825G

Only null left

Chemical Identifiers

CAS 6068-72-0
Molecular Formula C8H4ClNO
Molecular Weight (g/mol) 165.58
MDL Number MFCD00001822
InChI Key USEDMAWWQDFMFY-UHFFFAOYSA-N
Synonym 4-cyanobenzoylchloride, 4-cyano-benzoyl chloride, p-cyanobenzoyl chloride, unii-2577fde7a8, benzoyl chloride, 4-cyano, 4cyanobenzoyl chloride, 4-cyano-benzoylchloride, 4-cyanobenzoyl-chloride, 4-cyanpbenzoyl chloride, 4-cyano benzoylchloride
PubChem CID 80172
IUPAC Name 4-cyanobenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 110°C
Quantity 5 g
UN Number 3261
Formula Weight 165.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzoyl Chloride