missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoyl chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 6068-72-0
Molecular Formula C8H4ClNO
Molecular Weight (g/mol) 165.58
MDL Number MFCD00001822
InChI Key USEDMAWWQDFMFY-UHFFFAOYSA-N
Synonym 4-cyanobenzoylchloride, 4-cyano-benzoyl chloride, p-cyanobenzoyl chloride, unii-2577fde7a8, benzoyl chloride, 4-cyano, 4cyanobenzoyl chloride, 4-cyano-benzoylchloride, 4-cyanobenzoyl-chloride, 4-cyanpbenzoyl chloride, 4-cyano benzoylchloride
PubChem CID 80172
IUPAC Name 4-cyanobenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154550050
SDS View Documents
Thermo Scientific Chemicals
154550050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC154550250
SDS View Documents
Thermo Scientific Chemicals
154550250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6068-72-0
165.58
USEDMAWWQDFMFY-UHFFFAOYSA-N
80172
ClC(=O)C1=CC=C(C=C1)C#N
C8H4ClNO
MFCD00001822
4-cyanobenzoylchloride, 4-cyano-benzoyl chloride, p-cyanobenzoyl chloride, unii-2577fde7a8, benzoyl chloride, 4-cyano, 4cyanobenzoyl chloride, 4-cyano-benzoylchloride, 4-cyanobenzoyl-chloride, 4-cyanpbenzoyl chloride, 4-cyano benzoylchloride
4-cyanobenzoyl chloride

Specifications

6068-72-0
100.0
98%
C8H4ClNO
MFCD00001822
4-cyanobenzoylchloride, 4-cyano-benzoyl chloride, p-cyanobenzoyl chloride, unii-2577fde7a8, benzoyl chloride, 4-cyano, 4cyanobenzoyl chloride, 4-cyano-benzoylchloride, 4-cyanobenzoyl-chloride, 4-cyanpbenzoyl chloride, 4-cyano benzoylchloride
ClC(=O)C1=CC=C(C=C1)C#N
165.58
165.58
4-Cyanobenzoyl chloride, 98%
97.5
Authentic
Glass bottle
NCC6H4COCl
5g
USEDMAWWQDFMFY-UHFFFAOYSA-N
4-cyanobenzoyl chloride
80172
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 228-005-5

SDS
Documents

Documents

RUO – Research Use Only