missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. C18131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18131G 1 g
C18135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18131G Supplier TCI America Supplier No. C18131G
Only null left

Chemical Identifiers

CAS 49584-26-1
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.62
MDL Number MFCD00052337
InChI Key DBMFYTQPPBBKHI-UHFFFAOYSA-N
Synonym 4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
PubChem CID 39497
IUPAC Name 4-cyanobenzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
UN Number 2928
Formula Weight 201.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzenesulfonyl Chloride