missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanobenzenesulfonyl chloride, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 107°C to 110°C |
| Quantity | 1 g |
| UN Number | UN3261 |
| Beilstein | 777939 |
| Sensitivity | Moisture sensitive |
| IUPAC Name | 4-cyanobenzene-1-sulfonyl chloride |
| Formula Weight | 201.63 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Cyanobenzenesulfonyl chloride |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only